  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Beta Radiation Safety Racks"
https://tr.vwr.com/stibo/smallweb/std.lang.all/49/39/18944939.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"3"
Sort by:
Catalog Number: (113-8978)
Supplier:  VWR Collection
Description:   Rack, beta radiation safety, Microtube, VWR®, Acrylic, Thickness: 10 mm, For tubes: 1,5 ml, No. of holes: 4×6, 171×118×44 mm
UOM:  1 * 1 items
Supplier:  CBS / SCIENTIFIC
Description:   Rack, Beta Radiation Safety , Eppendorf tubes, Acrylic, For tubes: 1,5 ml, No. of holes: 12, 133×78×114 mm
Catalog Number: (CBSCRK-080)
Supplier:  CBS / SCIENTIFIC
Description:   Acrylic, radiation container with hinged lid.
UOM:  1 * 1 items
1 - 3  of 3