  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Radiation Shields"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/97/01/18989701.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"7"
Sort by:
Supplier:  VWR Collection
Description:   Beta radiation shield, VWR®, Acrylic, Angled, Transparent, Thickness: 10 mm, W×D×H: 304×150×450 mm
Catalog Number: (AZLOSRS001)
Supplier:  SCILABWARE
Description:   Acrylic protection shield; transparent, thickness: 10 mm.
UOM:  1 * 1 items
Supplier:  CBS / SCIENTIFIC
Description:   Shield beta radiation 45° angle with side cutout for easier access (35,4x54x26,1cm) 10mm thick 1 * 1 items
Catalog Number: (CBSCRL-024B)
Supplier:  CBS / SCIENTIFIC
Description:   Acrylic carrier shield, transparent, thickness: 12,7 mm.
UOM:  1 * 1 items
Supplier:  NALGE
Description:   Beta radiation shield, Nalgene®, Acrylic, Angled, Transparent, Thickness: 9 mm, W×D×H: 305×305×457 mm
Catalog Number: (KART572)
Supplier:  KARTELL
Description:   SAFETY SHIELD 305XH457X5MM BETA 1 * 1 items
UOM:  1 * 1 items
Catalog Number: (BELAF249760003)
Supplier:  Bel-Art
Description:   Acrylic protection shield; transparent, thickness 9,5 mm
UOM:  1 * 1 items
1 - 7  of 7