  • Criteria
    • In Stock (1)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Bioassay Dishes"
https://tr.vwr.com/stibo/smallweb/std.lang.all/68/35/20236835.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"5"
Sort by:
Supplier:  CORNING SCIENCE PRODUCTS
Description:   BioAssay dishes, 245×245 mm square
Supplier:  NUNC (INC)
Description:   Bioassay dishes, Nunc™
Supplier:  NUNC (INC)
Description:   Bioassay dishes, 4-well, for in vitro fertilisation, Nunc™
Catalog Number: (734-1727)
Supplier:  CORNING SCIENCE PRODUCTS
Description:   Bioassay dish, treated
UOM:  1 * 16 items
Catalog Number: (734-2096)
Supplier:  NUNC (INC)
Description:   Bioassay dish, Nunclon™Δ
UOM:  1 * 16 items
1 - 5  of 5