  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "CO2 and Temperature Monitoring Systems"
https://tr.vwr.com/stibo/smallweb/std.lang.all/93/24/18989324.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"5"
Sort by:
Supplier:  THERMO BRAND
Description:   Ambient measurements or monitoring of laboratory equipment.
Supplier:  EBRO
Description:   Software Winlog.Pro, for: Wireless multi-channel temperature monitoring systems, EBI 40
Catalog Number: (620-2452)
Supplier:  EBRO
Description:   For monitoring the CO2 concentration, the ambient temperature and the relative humidity.
UOM:  1 * 1 items
Supplier:  EBRO
Description:   The EBI 40 loggers are designed for mapping applications at high temperatures. In combination with thermal insulation box (620-1804), it can be used f...
Supplier:  THERMO BRAND
Description:   Power supply, 100/240v to 6 VDC, For: Smart-Vue monitoring system
1 - 5  of 5