  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "TLC Sorbents"
SearchPresentationType-VERTICAL
SearchResultCount:"6"
Sort by:
Supplier:  Merck
Description:   Selection of standardised and modified silica gels that are available in many granulations and supplied from one single source, dedicated to chromatog...
Catalog Number: (1.02330.0500)
Supplier:  Merck
Description:   Cellulose microcrystalline, <20 µm, Packed: In plastic
UOM:  1 * 500 g
Supplier:  ALFA (AESAR)
Description:   This TLC high purity grade silica gel without binder is used for thin layer chromatography plate preparation.
SDS
Supplier:  ALFA (AESAR)
Description:   This TLC high purity grade silica gel with gypsum binder is used for thin layer chromatography plate preparation.
SDS
Supplier:  ALFA (AESAR)
Description:   This TLC high purity grade silica gel without binder, with fluorescent indicator is used for thin layer chromatography plate preparation.
SDS
Supplier:  ALFA (AESAR)
Description:   This TLC high purity grade silica gel with gypsum binder and fluorescent indicator is used for thin layer chromatography plate preparation.
SDS
1 - 6  of 6