  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Gas Inlet Tubes"
https://tr.vwr.com/stibo/smallweb/std.lang.all/93/63/18989363.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"5"
Sort by:
Supplier:  BOHLENDER
Description:   Gas inlet tubes
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Gas inlet/air bleed tubes
Catalog Number: (201-2610)
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   Air bleeds
UOM:  1 * 1 items
Supplier:  SCILABWARE
Description:   Air/steam inlet tubes, straight, Quickfit®
Supplier:  SCILABWARE
Description:   Tubes, air/steam inlet, 90° bend, Quickfit®
1 - 5  of 5