  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Iodine Flasks"
https://tr.vwr.com/stibo/smallweb/std.lang.all/53/19/18945319.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"6"
Sort by:
Supplier:  DURAN GROUP GMBH
Description:   Flask, iodine determination,capacity 500ml, NS 29/32, with hollow, flat stopper,DURAN. 1 * 10 items
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   N/A
UOM:  1 * 1 items
Supplier:  LENZ LABORGLAS GMBH & CO.KG
Description:   N/A
Supplier:  WITEG LABORTECHNIK GMBH
Description:   FLASK JODINE, DURAN, WITH FUNNEL, HOLLOW NS-GLASS-STOPPER, EXTRA LONG STEM, 500ML NS29/32 1 * 1 items
UOM:  1 * 1 items
Supplier:  SCILABWARE
Description:   FLASK IODINE BOROSILICATE 500ML 1 * 10 items
Catalog Number: (KIML27200-250)
Supplier:  KIMBLE/KONTES
Description:   Flasks, manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements are us...
UOM:  1 * 8 items
1 - 6  of 6