  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Sulfonation Flasks"
https://tr.vwr.com/stibo/smallweb/std.lang.all/98/67/18989867.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"2"
Sort by:
Supplier:  WITEG LABORTECHNIK GMBH
Description:   Sulfonation flasks, Borosilicate glass, with three necks, 350 ml, Centre neck: 29/32 NS, Side necks: 14/23 NS
Supplier:  WITEG LABORTECHNIK GMBH
Description:   Sulfonation flasks, Borosilicate glass 3.3, with four necks, 500 ml, Centre neck: 45/40 NS, Side necks: 29/32 NS
1 - 2  of 2