  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "fraction-collection-tubes"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  CHROMACOL
Description:   Fraction collection tubes, Chromacol™, Rimless, round bottom, Nominal volume: 6 ml, Height: 75 mm, Ø ext.: 12 mm
1 - 1  of 1