  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "reverse-transcriptases"
https://tr-bot.vwr.com/stibo/smallweb/std.lang.all/97/39/18989739.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"15"
Sort by:
Catalog Number: (1B1326-50RXN)
Supplier:  VWR Chemicals
Description:   Ready One-Step RT-PCR Kit offers a single step procedure for performing RT-PCR reactions. Subsequent loading and visualisation of RT-PCR products are ...
UOM:  1 * 1 KIT
Supplier:  VWR Peqlab
Description:   peqGOLD M-MuLV H Plus Reverse Transcriptase (RT) is a genetically modified M-MuLV RT with the highest activity at 42°C. The enzyme possesses RNA-depen...
UOM:  1 * 10.000 EU
Supplier:  VWR Peqlab
Description:   peqGOLD M-MuLV H Minus Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. The enzyme posesses RNA-dependent and DNA dependent polymerase ...
UOM:  1 * 10.000 items
Supplier:  VWR Peqlab
Description:   The cDNA Synthesis Kit H Plus contains peqGOLD M-MuLV H Plus Reverse Transcriptase (RT). The genetically modified RT shows highest activity at 42 °C. ...
Catalog Number: (732-3271)
Supplier:  VWR Peqlab
Description:   The cDNA Synthesis Kit H Minus contains peqGOLD M-MuLV H Minus Reverse Transcriptase (RT). The genetically modified M-MuLV RT posesses RNA-dependent a...
UOM:  1 * 100 Tests
Catalog Number: (QUNT95143-020)
Supplier:  Quantabio
Description:   The qScript™ XLT One-Step RT-PCR Kit is a convenient and highly sensitive system for amplification of RNA templates up to 2 kb. cDNA synthesis and PCR...
UOM:  1 * 20 Assays
Supplier:  Quantabio
Description:   qScript™ cDNA Synthesis Kit contains qScript™ Reverse Transcriptase, a mixture of a MMLV reverse transcriptase derivative and ribonuclease inhibitor p...
Supplier:  Thermo Scientific
Description:   Maxima™ H Minus First Strand cDNA Synthesis Kit is a complete system for highly efficient synthesis of first strand cDNA. The kit uses the Maxima™ H M...
1 - 8  of 15
  1  2  Next