  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Dial Thermometers"
https://tr.vwr.com/stibo/smallweb/std.lang.all/93/43/18989343.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"8"
Sort by:
Supplier:  VWR Collection
Description:   Thermometer, 0 to 50c 1 * 1 items
Catalog Number: (620-0920)
Supplier:  VWR Collection
Description:   N/A
UOM:  1 * 1 items
Supplier:  VWR Collection
Description:   This thermometer with analog dial indicates freezer, refrigerator, or spoilage temperatures.
UOM:  1 * 1 items
Catalog Number: (AMART110410)
Supplier:  AMARELL, ARNO
Description:   Bimetal dial thermometers made of brass. The scale cover made of glass is used in heating systems and indutrial plants.
UOM:  1 * 1 items
Catalog Number: (AMARZ640870)
Supplier:  AMARELL, ARNO
Description:   Thermometer, for swimming pool, red filling, measuring range:0:40°C, diameter:190mm. 1 * 1 items
UOM:  1 * 1 items
Supplier:  Bel-Art
Description:   Thermometer, -20/150°C, maximum registering, autoclave, Bi-metal, with calibration certificate. 1 * 1 items
UOM:  1 * 1 items
Supplier:  AMARELL, ARNO
Description:   N/A
Catalog Number: (HACH4565500)
Supplier:  Hach
Description:   Thermometer for COD Reactor, dual gauge 0 - 200°C ± 1%, pk/1, divisions 2°C, Used with HACH COD Reactor 4560000 1 * 1 items
UOM:  1 * 1 items
1 - 8  of 8