  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "Digestion Tubes"
https://tr.vwr.com/stibo/smallweb/std.lang.all/93/45/18989345.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"2"
Sort by:
Supplier:  SCILABWARE
Description:   Digestion tubes, PYREX®, Borosilicate glass 3.3, with socket, Capacity: 75 ml, Length: 255 mm, Ø ext.: 26 mm
Supplier:  SCILABWARE
Description:   N/A
1 - 2  of 2