Would you like to log in again?
Thin Layer Chromatography
Thin Layer Chromatography
Thin Layer Chromatography (TLC) and High Performance Thin Layer Chromatography (HPTLC) are often referred to as Planar Chromatography. This is a modern classical separation technique, established worldwide and distinguished by flexibility, reliability and cost efficiency.
 Thin Layer Chromatography offers:
  • Enormous flexibility
  • Low cost separation method without the need for sophisticated instruments (instrumentation is required for HPTLC)
  • Multiple samples (up to 72) can be run simultaneously under identical conditions
  • Unsurpassed clarity and simultaneous visual evaluation of all samples and sample components 
  • Simplified sample preparation due to single use of the stationary phase