Would you like to log in again?
Cleanroom Wipes and Swabs
Wipes and Swabs
Cleanroom wipes are amongst the most important consumables in the cleanroom and controlled environments. They are integral to a complex cleaning process which removes contaminants generally invisible to the naked eye and support the maintenance of good basic hygiene together with a clean visual impression.
To ensure efficient cleaning, the cleanroom wipes must be able to absorb a wide range of surface contaminants in the shortest possible time and without themselves distributing further pollution in the form of particles, fibres, ions or microbiological contaminants.
Because cleanroom requirements vary widely, the selected wipes must be appropriate to the application. The various materials and structures of the wipes have a major effect on their technical and functional performance.
Certified Quality
Wipes from our partner ITW are supported by the most advanced quality control standards in the industry.
ITW Texwipe is the only manufacturer of wipes for use in ISO Class 5 or cleaner environments that is ISO certified.
basan Wipe Plans
We support you in the choice of suitable wipes for your application.