???store.login.keepSessionOpen???
???common.endingIn??? 
???store.common.expirationMessageSoft???
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microplate readers, Multiskan™ GO

Supplier: Thermo Scientific

The Multiskan™ GO is a compact, reliable, absorbance spectrophotometer microplate reader available in two instrument models, with or without a cuvette port. The instrument can be operated as a stand-alone unit using straightforward internal software or by using Thermo Scientific SkanIt™ software for PC control. Both the internal and PC software are available in multiple language versions, including English, French, German, Spanish, Portuguese, Russian, Chinese and Japanese. A broad wavelength range, including UV, makes this microplate reader ideal for a wide range of photometric applications, such as nucleic acid and protein analysis, enzyme assays, cytotoxicity and cell proliferation assays as well as apoptosis assays. The cuvette option provides a quick and easy way of measuring when there are only a couple of samples to be analysed.

  • Freely selectable wavelengths by the monochromator for the demands of various assays
  • Fast plate measurement and a full spectrum of a sample in less than 10 seconds
  • Both microplate and cuvette reading capability available
  • Shaking and incubation up to 45 °C for temperature-critical applications
  • Robotic compatibility for high throughput environments
The SkanIt™ software package supplied with the instruments provides easy assay optimisation, flexible data handling and convenient report formatting. The software also has a special remote control interface, which enables easy integration with automated systems.

µDrop™ Plate is a quick and easy tool for DNA and RNA assays using photometric measurement and sample volumes down to 2 µl. The fixed light path of the µDrop™ Plate allows direct calculation of the nucleic acid concentrations of the samples.

dsDNA detection range from a few micrograms to a few milligrams per millilitre
Sixteen sample positions for quick and easy measurement of sample volumes down to 2 µl
Compatible with an eight channel pipette for easy sample addition
Quick and easy to wipe off the samples in serial measurements
Format flexibility with measurement slot for a rectangular cuvette (10 mm path length)
Compatible with Multiskan™ GO and Varioskan™ Flash



Log in to see your contract pricing and availability.
  Remember me on this device.
 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Multiskan™ GO with cuvette port 1 736-0560
Multiskan™ GO without cuvette port 1 736-0559

µDrop Plate
Description Pk VWR Catalog Number Quantity
µDrop™ Plate for up 16 samples, volumes 2 - 10µL, for use with microplate readers Multiskan™ GO and Varioskan™ Flash 1 736-0754
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model Multiskan™ GO with cuvette port Multiskan™ GO without cuvette port
Light source Xenon flash lamp
Wavelength range (nm) 200 - 1000
Wavelength selection Monochromator
Bandwidth <2,5 nm
Plate types 96- and 384-well
Absorbance range 0 - 4 Abs
Resolution 1 nm, 0,0001 Abs
Absorbance accuracy ±1,0% or ±0,003 Abs (0 - 2 Abs), ±2,0% (2 - 2,5 Abs)
Precision SD <0,003 Abs or CV <0,5% (cuvettes) SD <0,003 Abs or CV <0,5% (Precision mode)
SD <0,003 Abs or CV <1,0% (Fast mode)
Cuvette type (mm) W×D×H: 12,5×12,5×(44 - 58) -
Linearity (96-well plate) 0 - 2,5 Abs, ±2% at 450 nm (also cuvette with beam window ≥2 mm ) 0 - 2,5 Abs, ±2% at 450 nm
Measurement time (96-well plate) 6 s (from A1 back to A1)
Measurement time (384-well plate) 10 s (from A1 back to A1)
Display 4,5 inch colour display
Scan speed 200 - 1000 nm in 10 seconds
Software Internal software or PC control with SkanIt™ software
Shaking Linear (plates) Linear
Incubation range (°C) From ambient +2...45
Interfaces USB for computer connection
USB memory stick for data export
USB for external printer (HP PCL5 type)
W×D×H (mm) 285×430×260
Weight (kg) 10,8
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
???store.checkout.costsavings.reviewAndCompare???
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty