Would you like to log in again?
Acetonitrile, anhydrous (max. 0.003% H₂O) HiPerSolv CHROMANORM® Reag. Ph. Eur., USP, ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
(Cyanomethane Ethanenitrile Methyl cyanide Ethanoic acid nitrile)
R: 11-20/21/22-36
S: 16-36/37
H: H225 H302+H312+H332 H319
P: P210 P280 P305+P351+P338 P309+P310
Danger
  Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: -45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
MDL Number: MFCD00001878
CAS Number: 75-05-8
ADR: 3,II
REACH: 01-2119471307-38
https://tr.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Specification Test Results
Assay Min. 99.95 %
Solution (100 g/l) Neutral
Acidity Max. 0.0008 meq/g
Alkalinity Max. 0.0006 meq/g
Boiling point 80 - 82 °C
Colouration Max. 10 APHA
Density (20/4) 0.781 - 0.784
Density (20/20) 0.782 - 0.785
n 20/D 1.343 - 1.345
Evaporation residue Max. 2 ppm
Water Max. 30 ppm
Absorbance (190 nm) Max. 1
Absorbance (195 nm) Max. 0.1
Absorbance (200 nm) Max. 0.02
Absorbance (210 nm) Max. 0.02
Absorbance (230 nm) Max. 0.01
Absorbance (220 nm) Max. 0.01
Absorbance (250 nm) Max. 0.005
Absorbance (240 nm) Max. 0.005
Transmittance (190 nm) Min. 10 %
Absorbance (240-420 nm) Max. 0.005
Transmittance (200 nm) Min. 95 %
Transmittance (195 nm) Min. 80 %
Transmittance (220 nm) Min. 97 %
Transmittance (210 nm) Min. 96 %
Transmittance (240 nm) Min. 99 %
Transmittance (230 nm) Min. 98 %
Transmittance (240-420 nm) Min. 99 %
Transmittance (250 nm) Min. 99 %
Gradient grade (210 nm) Max. 3 mAU
Fluorescence (as quinine; 254 nm) Max. 1 ppb
Conforms to ACS (HPLC/UV) Passes test
Conforms to Reag. Ph.Eur. Passes test
Gradient (254 nm) Max. 1 mAU
Conforms to ACS Passes test
Conforms to USP Passes test
Log in to see your contract pricing and availability.
  Remember me on this device.
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Pack type Pk VWR Catalog Number Quantity
Glass bottle 2,5 l 83639.320
Get a discount if you order 4 or more. Login in now for details.
Glass bottle 1 l 83639.290
Get a discount if you order 6 or more. Login in now for details.
Aluminium bottle 5 l 83639.360
Get a discount if you order 4 or more. Login in now for details.
Aluminium bottle 10 l 83639.910
Glass bottle 4 l 83639.400
Aluminium bottle 28 l 83639.928
- 185 l 83639.500
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
???store.checkout.costsavings.reviewAndCompare???
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty