Would you like to log in again?
UV/Vis spectrophotometer, UV-6300PC
Supplier: VWR Collection
A double beam spectrophotometer suitable for a wide range of applications including general research, pharmaceutical, biochemical and clinical laboratory use, new material development and quality control.
  • Standard photometrics - Absorption, transmission, concentration
  • Quantitative analysis and kinetics, multi-wavelength scan, spectrum scan
  • DNA/RNA and protein analysis
The instrument can be used in stand-alone mode or in conjunction with a PC using the supplied UV-Vis Analyst software which then delivers more powerful data processing, expanded data collecting, and removes the limit on storage capability.
Delivery information: Supplied with four optical glass cells, two quartz cells, dust cover, USB cable, USB (8 GB) stick, UV-Vis Analyst software for PC control, cell holders for sample and reference, power cables with UK, CH and EU plugs.
Log in to see your contract pricing and availability.
  Remember me on this device.
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Model Pk VWR Catalog Number Quantity
UV-6300PC 1 634-6041
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Optical design Double beam
Wavelength range (nm) 190 - 1100
Wavelength accuracy (nm) ±0,3
Display LCD (320×240 P)
Photometric range −0,3 to +3,0 A; 0 to 200% T; 0 - 9999 Conc
Photometric accuracy ±0,002 A @ 1 A ; ≤0,3% T
Photometric stability 0,001 A/h @ 500 nm
Light source Deuterium and tungsten halogen
Stray light ≤0,05% T @ 220 and 360 nm
Scan speed (nm/min) 100 - 3000
Baseline flatness 0,001 A (200 - 1000 nm)
Scan Intervals: 0,1; 0,2; 0,5; 1,0; 5,0 nm
PC software UV-Vis Analyst
Control Internal control via keypad (optional PC control)
Storage capacity 200 results and 200 curves
Interfaces USB and parallel printer interface
Languages English, French, German, Spanish
Weight (kg) 26
W×D×H (mm) 590×420×260
ACCESSORIES
Expand All / Collapse All
Accessories
Description For Pk VWR Catalog Number Quantity
Cell holder (sample), 1 position, for 10 mm path length standard cells UV-6300PC 1 634-0777
Cell holder (reference), 1 position, for 10 mm path length standard cells UV-6300PC 1 634-0778
Cell holder for microcells with beam height 15 mm V-1200, UV-1600PC, V-3000PC, UV-3100PC, UV-6300PC 1 634-0687
Cell holder, 4 position, for rectangular cells 10 to 50 mm path length UV-6300PC 1 634-6045
Cell holder, 4-position, for rectangular cells with 100 mm path length UV-6300PC 1 634-6046
Cell holder, single position, for rectangular cells with 100 mm path length UV-6300PC 1 634-6047
Cell holder, single position, for cylindrical cells UV-6300PC 1 634-6048
Single position cell holder for 10 mm path length, standard cells (external water bath required) UV-6300PC 1 634-6049
Four-position cell holder for 10 mm path length, standard cells (external water bath required) - 1 634-6050
Cell holder, single position, for test tubes, max. height 100 mm UV-6300PC 1 634-6051
Cell holder, 8-position auto cell changer UV-1600PC, V-3000PC, UV-3100PC, UV-6300PC 1 634-6010
Cell holder, for solid samples, max. thickness 3,5 mm UV-6300PC 1 634-6052
A-100, constant temperature (Peltier) sipper system, beam height 15 mm, (appropriate flow cell required) UV-1600PC, V-3000PC, UV-3100PC, UV-6300PC 1 634-0763
A-101 sipper system with beam height 15 mm, (appropriate flow cell required) UV-1600PC, V-3000PC, UV-3100PC, UV-6300PC 1 634-0699
A-102, constant temperature (Peltier) system with beam height 15 mm UV-1600PC, V-3000PC, UV-3100PC, UV-6300PC 1 634-0698
Thermal printer V-1200, UV-1600PC, V-3000PC, UV-3100PC, UV-6300PC 1 634-6039
Lamp, deuterium UV-1600PC, UV-3100PC, UV-6300PC 1 634-6038
Dust cover UV-6300PC 1 634-6044
Cuvette, quartz, square, 10 mm path length UV-1600PC, UV-3100PC, UV-6300PC 2 634-6018
Halogen lamp (12 V/20 W), Osram - 1 634-0776
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
???store.checkout.costsavings.reviewAndCompare???
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty